Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NECAP2
Entrez Gene ID
55707
Gene Name
NECAP endocytosis associated 2
Gene Aliases
-
Chromosome Location
Chr.1: 16440672 - 16460090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145277.1 NP_001138749.1 1 - 2 187 119
NM_018090.4 NP_060560.1 1 - 2 187 119
XM_017001721.1 XP_016857210.1 1 - 2 135 119
GenBank mRNA AK001282.1 - 1 - 2 119 119
AK023545.1 - 1 - 2 123 119
AK024468.1 - 1 - 2 101 119
AK223344.1 - 1 - 2 135 119
AK300764.1 - 1 - 2 133 119
BC017014.1 - 1 - 2 118 119
BC018914.2 - 1 - 2 121 119
BM751287.1 - 1 - 2 187 119

View Full Product Details