The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018112.2 | NP_060582.1 | 1 - 2 | 249 | 55 |
XM_005252076.2 | XP_005252133.1 | 1 - 2 | 230 | 55 | |
XM_005252077.3 | XP_005252134.1 | 1 - 2 | 230 | 55 | |
XM_011518829.1 | XP_011517131.1 | 1 - 2 | 230 | 55 | |
XM_011518831.2 | XP_011517133.1 | 1 - 2 | 230 | 55 | |
XM_011518832.2 | XP_011517134.1 | 1 - 2 | 242 | 55 | |
XM_011518833.2 | XP_011517135.1 | 1 - 2 | 241 | 55 | |
GenBank mRNA | AK001355.1 | - | 1 - 2 | 230 | 55 |
BC000049.2 | - | 1 - 2 | 205 | 55 |