The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018169.3 | NP_060639.3 | 4 - 5 | 5414 | 116 |
XM_005253405.2 | XP_005253462.1 | 3 - 4 | 5865 | 116 | |
XM_011520722.1 | XP_011519024.1 | 4 - 5 | 5574 | 116 | |
XM_017019550.1 | XP_016875039.1 | 3 - 4 | 6274 | 116 | |
GenBank mRNA | AK001514.1 | - | 1 - 2 | 898 | 116 |
BC098115.1 | - | 1 - 2 | 878 | 116 | |
BC114509.1 | - | 1 - 2 | 878 | 116 | |
BC114956.1 | - | 1 - 2 | 878 | 116 | |
BC140819.1 | - | 4 - 5 | 5365 | 116 |