The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018227.5 | NP_060697.4 | 25 - 26 | 2313 | 71 |
XM_017008359.1 | XP_016863848.1 | 25 - 26 | 2290 | 71 | |
GenBank mRNA | AB014773.1 | - | 25 - 26 | 2261 | 71 |
AK001670.1 | - | 7 - 8 | 662 | 71 | |
AK094164.1 | - | 17 - 18 | 1519 | 71 | |
AK314371.1 | - | 25 - 26 | 2290 | 71 | |
AL832015.2 | - | 25 - 26 | 2282 | 71 | |
AL832458.2 | - | 25 - 26 | 2282 | 71 | |
AY359880.1 | - | 12 - 13 | 1634 | 71 | |
BC126484.1 | - | 25 - 26 | 2308 | 71 | |
BC126486.1 | - | 25 - 26 | 2308 | 71 | |
EF623993.1 | - | 25 - 26 | 2261 | 71 |