The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018959.3 | NP_061832.2 | 1 - 2 | 233 | 61 |
NM_170711.2 | NP_733829.1 | 1 - 2 | 233 | 61 | |
XM_005259531.3 | XP_005259588.1 | 1 - 2 | 186 | 61 | |
XM_005259534.3 | XP_005259591.1 | 1 - 2 | 192 | 61 | |
XM_005259535.3 | XP_005259592.1 | 1 - 2 | 194 | 61 | |
XM_005259536.3 | XP_005259593.1 | 1 - 2 | 194 | 61 | |
XM_017026581.1 | XP_016882070.1 | 1 - 2 | 228 | 61 | |
GenBank mRNA | AF181719.1 | - | 1 - 2 | 136 | 61 |
AL360261.1 | - | 1 - 2 | 128 | 61 | |
BC012062.1 | - | 1 - 2 | 101 | 61 | |
BI909277.1 | - | 1 - 2 | 217 | 61 | |
BU557183.1 | - | - | 233 | 61 |