The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003692.1 | NP_001003692.1 | 2 - 3 | 267 | 75 |
NM_001318129.1 | NP_001305058.1 | 2 - 3 | 221 | 75 | |
NM_019103.2 | NP_061976.1 | 3 - 4 | 382 | 75 | |
GenBank mRNA | AA312132.1 | - | 2 - 3 | 267 | 75 |
AK292671.1 | - | 2 - 3 | 248 | 75 | |
AL160311.1 | - | 2 - 3 | 266 | 75 | |
BC009717.1 | - | 2 - 3 | 232 | 75 | |
BG285784.1 | - | 3 - 4 | 346 | 75 | |
BQ931527.1 | - | 2 - 3 | 195 | 75 | |
BU598307.1 | - | 2 - 3 | 269 | 75 | |
CR456353.1 | - | 1 - 2 | 154 | 75 | |
CR457393.1 | - | 1 - 2 | 131 | 75 | |
DT217179.1 | - | 3 - 4 | 398 | 75 |