The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001503.1 | NP_001001503.1 | 1 - 2 | 113 | 74 |
NM_021075.3 | NP_066553.3 | 1 - 2 | 113 | 74 | |
XM_011529586.2 | XP_011527888.1 | 1 - 2 | 110 | 74 | |
XM_017028359.1 | XP_016883848.1 | 1 - 2 | 108 | 74 | |
GenBank mRNA | AK289586.1 | - | 1 - 2 | 67 | 74 |
AK308228.1 | - | 1 - 2 | 70 | 74 | |
BC021217.2 | - | 1 - 2 | 100 | 74 | |
BC033766.1 | - | 1 - 2 | 71 | 74 | |
BC054016.1 | - | 1 - 2 | 114 | 74 | |
BF311986.1 | - | 1 - 2 | 58 | 74 | |
BI829092.1 | - | 1 - 2 | 116 | 74 | |
CR542190.1 | - | 1 - 2 | 47 | 74 |