The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022147.2 | NP_071430.2 | 1 - 2 | 218 | 58 |
GenBank mRNA | AJ251832.1 | - | 1 - 2 | 210 | 58 |
AY562238.1 | - | 1 - 2 | 156 | 58 | |
BC013161.1 | - | 1 - 2 | 218 | 58 |