Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TINAGL1
Entrez Gene ID
64129
Gene Name
tubulointerstitial nephritis antigen like 1
Gene Aliases
ARG1, LCN7, LIECG3, TINAGRP
Chromosome Location
Chr.1: 31576384 - 31587686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204415.1 NP_001191344.1 2 - 3 107 61
NM_022164.2 NP_071447.1 3 - 4 499 61
XM_005271106.2 XP_005271163.1 3 - 4 499 61
XM_005271107.2 XP_005271164.1 2 - 3 144 61
GenBank mRNA AB050716.1 - 3 - 4 449 61
AF205436.1 - 3 - 4 425 61
AF236150.1 - 3 - 4 427 61
AK074124.1 - 2 - 3 488 61
AK075398.1 - 3 - 4 472 61
AK292770.1 - 3 - 4 473 61
AK301070.1 - 3 - 4 509 61
AY358421.1 - 3 - 4 472 61
BC009048.1 - 1 - 2 30 61
BC064633.1 - 3 - 4 457 61
BM923842.1 - 2 - 3 107 61
DA656570.1 - 3 - 4 501 61

View Full Product Details