Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC1
Entrez Gene ID
64215
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Gene Aliases
DNAJL1, ERdj1, HTJ1, MTJ1
Chromosome Location
Chr.10: 21756548 - 22003768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022365.3 NP_071760.2 10 - 11 1434 70
GenBank mRNA AF427142.1 - 3 - 4 310 70
AK026062.1 - 7 - 8 707 70
AK027263.1 - 10 - 11 1254 70
AY225122.1 - 10 - 11 1147 70
BC030955.1 - 6 - 7 617 70
BC110894.1 - 10 - 11 1434 70
BC139752.1 - 2 - 3 154 70

View Full Product Details