The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022480.3 | NP_071925.2 | 2 - 3 | 2003 | 85 |
GenBank mRNA | AK022649.1 | - | 1 - 2 | 1424 | 85 |
AK056663.1 | - | 3 - 4 | 1845 | 85 | |
AK092231.1 | - | 2 - 3 | 1965 | 85 | |
AL136796.1 | - | 2 - 3 | 1915 | 85 | |
AY764034.1 | - | 2 - 3 | 1985 | 85 | |
BC028100.1 | - | 2 - 3 | 1922 | 85 |