The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022735.3 | NP_073572.2 | 5 - 6 | 948 | 87 |
GenBank mRNA | AB043587.2 | - | 5 - 6 | 956 | 87 |
AK025520.1 | - | 5 - 6 | 949 | 87 | |
AK314468.1 | - | 5 - 6 | 949 | 87 | |
AY150218.1 | - | 5 - 6 | 927 | 87 | |
BC034563.2 | - | 3 - 4 | 397 | 87 | |
BC045533.1 | - | 5 - 6 | 934 | 87 | |
BC060792.1 | - | 5 - 6 | 944 | 87 |