The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024097.3 | NP_077002.2 | 2 - 3 | 264 | 105 |
GenBank mRNA | BC001508.2 | - | 2 - 3 | 240 | 105 |
BC001711.1 | - | 2 - 3 | 240 | 105 | |
DA684965.1 | - | 2 - 3 | 264 | 105 |