The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024700.3 | NP_078976.2 | 2 - 3 | 446 | 103 |
GenBank mRNA | AK022615.1 | - | 2 - 3 | 404 | 103 |
AK027622.1 | - | 2 - 3 | 392 | 103 | |
AK293229.1 | - | 3 - 4 | 358 | 103 | |
AK300796.1 | - | 3 - 4 | 267 | 103 | |
AK315574.1 | - | 2 - 3 | 413 | 103 | |
AY081909.1 | - | 2 - 3 | 331 | 103 | |
BC027040.1 | - | 2 - 3 | 404 | 103 | |
BP364813.1 | - | 2 - 3 | 408 | 103 | |
CX759814.1 | - | 2 - 3 | 446 | 103 |