The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142886.1 | NP_001136358.1 | 5 - 6 | 786 | 80 |
NM_024794.2 | NP_079070.1 | 4 - 5 | 822 | 80 | |
GenBank mRNA | AK026061.1 | - | 4 - 5 | 822 | 80 |
BC115002.1 | - | 5 - 6 | 696 | 80 | |
BC132958.1 | - | 4 - 5 | 786 | 80 | |
BC132960.1 | - | 4 - 5 | 786 | 80 |