The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024807.3 | NP_079083.2 | 2 - 3 | 561 | 70 |
GenBank mRNA | AF534824.1 | - | 2 - 3 | 560 | 70 |
AK023755.1 | - | 2 - 3 | 560 | 70 | |
AY358171.1 | - | 2 - 3 | 562 | 70 | |
BC125079.1 | - | 2 - 3 | 560 | 70 |