The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024813.2 | NP_079089.2 | 6 - 7 | 596 | 70 |
XM_005271223.2 | XP_005271280.1 | 6 - 7 | 538 | 70 | |
XM_011542167.2 | XP_011540469.1 | 6 - 7 | 538 | 70 | |
XM_017002362.1 | XP_016857851.1 | 6 - 7 | 538 | 70 | |
XM_017002363.1 | XP_016857852.1 | 6 - 7 | 538 | 70 | |
XM_017002364.1 | XP_016857853.1 | 6 - 7 | 538 | 70 | |
GenBank mRNA | AK023212.1 | - | 6 - 7 | 513 | 70 |
AK225054.1 | - | 6 - 7 | 508 | 70 | |
BC031070.1 | - | 6 - 7 | 502 | 70 | |
BC039014.1 | - | 6 - 7 | 596 | 70 |