The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083600.2 | NP_001077069.1 | 1 - 2 | 199 | 75 |
NM_001317094.1 | NP_001304023.1 | 1 - 2 | 199 | 75 | |
NM_001317096.1 | NP_001304025.1 | 1 - 2 | 199 | 75 | |
NM_001317097.1 | NP_001304026.1 | 1 - 2 | 199 | 75 | |
NM_001317098.1 | NP_001304027.1 | 1 - 2 | 199 | 75 | |
NM_024845.3 | NP_079121.1 | 1 - 2 | 199 | 75 | |
GenBank mRNA | AK024216.1 | - | 1 - 2 | 106 | 75 |
AK130792.1 | - | 1 - 2 | 106 | 75 | |
AK225178.1 | - | 1 - 2 | 106 | 75 | |
AK298566.1 | - | 1 - 2 | 108 | 75 | |
AK302361.1 | - | 1 - 2 | 64 | 75 | |
AK311524.1 | - | 1 - 2 | 106 | 75 | |
AY358543.1 | - | 1 - 2 | 95 | 75 | |
BG822206.1 | - | 1 - 2 | 55 | 75 | |
DA736660.1 | - | 1 - 2 | 192 | 75 | |
DC339939.1 | - | 1 - 2 | 199 | 75 |