The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 9 - 10 | 1404 | 65 |
XM_005270185.4 | XP_005270242.2 | 9 - 10 | 1435 | 65 | |
XM_011540213.1 | XP_011538515.1 | 9 - 10 | 1435 | 65 | |
XM_017016696.1 | XP_016872185.1 | 9 - 10 | 1372 | 65 | |
XM_017016697.1 | XP_016872186.1 | 9 - 10 | 1388 | 65 | |
XM_017016698.1 | XP_016872187.1 | 9 - 10 | 1099 | 65 | |
XM_017016699.1 | XP_016872188.1 | 9 - 10 | 1388 | 65 | |
XM_017016700.1 | XP_016872189.1 | 9 - 10 | 985 | 65 | |
XM_017016701.1 | XP_016872190.1 | 9 - 10 | 1435 | 65 | |
GenBank mRNA | AF264912.1 | - | 9 - 10 | 1107 | 65 |
AF309033.1 | - | 9 - 10 | 1358 | 65 | |
AF329696.1 | - | 9 - 10 | 1246 | 65 | |
AF342982.1 | - | 9 - 10 | 1107 | 65 | |
AF438201.1 | - | 9 - 10 | 1486 | 65 | |
AK314612.1 | - | 9 - 10 | 1372 | 65 |