Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOST
Entrez Gene ID
50964
Gene Name
sclerostin
Gene Aliases
CDD, DAND6, SOST1, VBCH
Chromosome Location
Chr.17: 43753731 - 43758788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025237.2 NP_079513.1 1 - 2 266 81
GenBank mRNA AF326739.1 - 1 - 2 266 81
AF331844.1 - 1 - 2 256 81
AY358627.1 - 1 - 2 280 81
BC101086.1 - 1 - 2 246 81
BC101087.1 - 1 - 2 246 81
BC101088.1 - 1 - 2 246 81
BC101089.1 - 1 - 2 246 81

View Full Product Details