Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STMN4
Entrez Gene ID
81551
Gene Name
stathmin 4
Gene Aliases
RB3
Chromosome Location
Chr.8: 27235323 - 27258865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283054.1 NP_001269983.1 5 - 6 683 80
NM_030795.3 NP_110422.2 6 - 7 764 80
GenBank mRNA AJ303455.1 - 4 - 5 593 80
AK225142.1 - 5 - 6 649 80
AL136568.1 - 5 - 6 653 80
BC011520.1 - 6 - 7 711 80
BC111001.1 - 5 - 6 644 80

View Full Product Details