The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185096.1 | NP_001172025.1 | 2 - 3 | 250 | 80 |
NM_031426.3 | NP_113614.1 | 2 - 3 | 250 | 80 | |
NR_033701.1 | 2 - 3 | 272 | 80 | ||
GenBank mRNA | AK091912.1 | - | 2 - 3 | 187 | 80 |
AK314600.1 | - | 2 - 3 | 197 | 80 | |
AL136566.1 | - | 2 - 3 | 184 | 80 | |
AL833896.1 | - | 2 - 3 | 184 | 80 | |
AY359067.1 | - | 2 - 3 | 201 | 80 | |
BC021253.2 | - | 2 - 3 | 175 | 80 | |
BI766795.1 | - | 2 - 3 | 223 | 80 | |
DA285481.1 | - | 2 - 3 | 251 | 80 |