Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD99L2
Entrez Gene ID
83692
Gene Name
CD99 molecule like 2
Gene Aliases
CD99B, MIC2L1
Chromosome Location
Chr.X: 150766336 - 150898816 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184808.1 NP_001171737.1 6 - 7 667 85
NM_031462.3 NP_113650.2 9 - 10 886 85
NM_134445.3 NP_604394.1 6 - 7 670 85
NM_134446.3 NP_604395.1 7 - 8 739 85
GenBank mRNA AK023776.1 - 8 - 9 572 85
AK024278.1 - 5 - 6 393 85
AK290200.1 - 9 - 10 662 85
AK291375.1 - 7 - 8 552 85
AL136580.1 - 9 - 10 775 85
AY078165.1 - 9 - 10 694 85
AY078166.1 - 7 - 8 547 85
AY078167.1 - 6 - 7 478 85
AY358837.1 - 9 - 10 759 85
BC025729.1 - 10 - 11 2107 85
BC030536.1 - 9 - 10 746 85
KU178797.1 - 9 - 10 658 85
KU178798.1 - 3 - 4 229 85

View Full Product Details