The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018653.3 | NP_061123.3 | 1 - 2 | 64 | 92 |
GenBank mRNA | AJ276102.1 | - | 1 - 2 | 73 | 92 |