The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271867.1 | NP_001258796.1 | 8 - 9 | 658 | 96 |
NM_001271868.1 | NP_001258797.1 | 8 - 9 | 574 | 96 | |
NM_001516.4 | NP_001507.2 | 9 - 10 | 738 | 96 | |
XM_017019228.1 | XP_016874717.1 | 9 - 10 | 656 | 96 | |
GenBank mRNA | AK298128.1 | - | 8 - 9 | 568 | 96 |
AK313200.1 | - | 9 - 10 | 648 | 96 | |
BC031030.1 | - | 6 - 7 | 399 | 96 | |
BC039726.2 | - | 8 - 9 | 604 | 96 | |
BC047868.2 | - | 8 - 9 | 604 | 96 | |
BC065250.1 | - | 9 - 10 | 631 | 96 | |
DA953586.1 | - | 8 - 9 | 486 | 96 | |
Z30093.1 | - | 8 - 9 | 617 | 96 |