The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177716.1 | NP_001171187.1 | 2 - 3 | 332 | 83 |
NM_001177718.1 | NP_001171189.1 | 2 - 3 | 480 | 83 | |
NM_003597.4 | NP_003588.1 | 2 - 3 | 477 | 83 | |
GenBank mRNA | AF028008.1 | - | 2 - 3 | 332 | 83 |
AF272830.1 | - | 2 - 3 | 451 | 83 | |
AJ275989.1 | - | 2 - 3 | 315 | 83 | |
AK289816.1 | - | 2 - 3 | 406 | 83 | |
AK315345.1 | - | 2 - 3 | 457 | 83 | |
AK316439.1 | - | 2 - 3 | 332 | 83 | |
BC063286.1 | - | 2 - 3 | 477 | 83 | |
BC069383.1 | - | 2 - 3 | 365 | 83 | |
BC074922.2 | - | 2 - 3 | 347 | 83 | |
BG701929.1 | - | 2 - 3 | 487 | 83 | |
CB990375.1 | - | 2 - 3 | 504 | 83 |