The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005955.2 | NP_005946.2 | 4 - 5 | 922 | 78 |
XM_011541491.2 | XP_011539793.1 | 4 - 5 | 1086 | 78 | |
XM_011541493.2 | XP_011539795.1 | 4 - 5 | 926 | 78 | |
XM_011541494.1 | XP_011539796.1 | 5 - 6 | 945 | 78 | |
GenBank mRNA | AK225923.1 | - | 4 - 5 | 894 | 78 |
AK309584.1 | - | 5 - 6 | 945 | 78 | |
AK314233.1 | - | 4 - 5 | 894 | 78 | |
BC014454.1 | - | 4 - 5 | 897 | 78 | |
X78710.1 | - | 4 - 5 | 864 | 78 |