The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301726.1 | NP_001288655.1 | 4 - 5 | 1331 | 65 |
NM_006602.3 | NP_006593.2 | 4 - 5 | 1334 | 65 | |
XM_005260185.3 | XP_005260242.1 | 4 - 5 | 1238 | 65 | |
XM_011528497.2 | XP_011526799.1 | 4 - 5 | 1238 | 65 | |
XM_011528498.2 | XP_011526800.1 | 4 - 5 | 1238 | 65 | |
XM_011528499.1 | XP_011526801.1 | 4 - 5 | 1238 | 65 | |
GenBank mRNA | AB012124.1 | - | 4 - 5 | 1195 | 65 |
AF070992.1 | - | 4 - 5 | 1172 | 65 | |
AJ271337.1 | - | 3 - 4 | 1043 | 65 | |
BC046933.1 | - | 4 - 5 | 1335 | 65 |