Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX1
Entrez Gene ID
8092
Gene Name
ALX homeobox 1
Gene Aliases
CART1, FND3, HEL23
Chromosome Location
Chr.12: 85280258 - 85301784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006982.2 NP_008913.2 1 - 2 232 92
GenBank mRNA BC010923.1 - 1 - 2 238 92
EU794597.1 - 1 - 2 238 92
U31986.1 - 1 - 2 383 92

View Full Product Details