The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291494.1 | NP_001278423.1 | 3 - 4 | 1012 | 89 |
NM_003638.2 | NP_003629.2 | 3 - 4 | 1012 | 89 | |
XM_011519752.2 | XP_011518054.1 | 3 - 4 | 1006 | 89 | |
GenBank mRNA | AK297740.1 | - | 3 - 4 | 524 | 89 |
AK308240.1 | - | 3 - 4 | 529 | 89 | |
L36531.1 | - | 3 - 4 | 334 | 89 |