Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRF1
Entrez Gene ID
3659
Gene Name
interferon regulatory factor 1
Gene Aliases
IRF-1, MAR
Chromosome Location
Chr.5: 132481609 - 132490789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002198.2 NP_002189.1 1 - 2 233 139
XM_011543378.2 XP_011541680.1 1 - 2 233 139
GenBank mRNA AK314025.1 - 1 - 2 233 139
BC009483.2 - 1 - 2 202 139
KC209827.1 - 1 - 2 115 139
KC209828.1 - 1 - 2 115 139
KJ200632.1 - 1 - 2 115 139
KJ200633.1 - 1 - 2 115 139
KJ200634.1 - 1 - 2 115 139
X14454.1 - 1 - 2 196 140

View Full Product Details