The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003037.3 | NP_003028.1 | 1 - 2 | 442 | 68 |
NR_104399.1 | 1 - 2 | 442 | 68 | ||
NR_104400.1 | 1 - 2 | 442 | 68 | ||
NR_104401.1 | 1 - 2 | 442 | 68 | ||
XM_005245456.3 | XP_005245513.1 | 1 - 2 | 668 | 68 | |
XM_011509905.2 | XP_011508207.1 | 1 - 2 | 668 | 68 | |
XM_017002129.1 | XP_016857618.1 | 1 - 2 | 668 | 68 | |
XM_017002130.1 | XP_016857619.1 | 1 - 2 | 668 | 68 | |
XM_017002131.1 | XP_016857620.1 | 1 - 2 | 668 | 68 | |
GenBank mRNA | AK304237.1 | - | 1 - 2 | 152 | 68 |
AK314244.1 | - | 1 - 2 | 386 | 68 | |
AY040554.1 | - | 1 - 2 | 76 | 68 | |
BC012602.1 | - | 1 - 2 | 429 | 68 | |
BC132792.1 | - | 1 - 2 | 108 | 68 | |
BC144498.1 | - | 1 - 2 | 108 | 68 | |
DB133023.1 | - | 1 - 2 | 422 | 68 | |
KF471075.1 | - | 1 - 2 | 89 | 68 | |
U33017.1 | - | 1 - 2 | 209 | 68 |