The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005856.2 | NP_005847.1 | 1 - 2 | 117 | 82 |
GenBank mRNA | AJ001016.1 | - | 1 - 2 | 85 | 82 |
AK222469.1 | - | 1 - 2 | 81 | 82 | |
AK311871.1 | - | 1 - 2 | 56 | 82 | |
AY265459.1 | - | 1 - 2 | 56 | 82 | |
BC022304.1 | - | 1 - 2 | 84 | 82 | |
BP347072.1 | - | 1 - 2 | 117 | 82 |