Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHP1
Entrez Gene ID
4867
Gene Name
nephrocystin 1
Gene Aliases
JBTS4, NPH1, SLSN1
Chromosome Location
Chr.2: 110123336 - 110205062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000272.3 NP_000263.2 12 - 13 1424 69
NM_001128178.1 NP_001121650.1 12 - 13 1256 69
NM_001128179.1 NP_001121651.1 10 - 11 1067 69
NM_207181.2 NP_997064.2 12 - 13 1421 69
XM_005263676.1 XP_005263733.1 12 - 13 1205 69
XM_005263677.1 XP_005263734.1 12 - 13 1202 69
XM_005263678.2 XP_005263735.1 12 - 13 1373 69
XM_005263679.1 XP_005263736.1 12 - 13 1202 69
XM_006712551.1 XP_006712614.1 12 - 13 1373 69
XM_011511244.1 XP_011509546.1 12 - 13 1373 69
XM_017004218.1 XP_016859707.1 12 - 13 1205 69
GenBank mRNA AF023674.1 - 12 - 13 1325 69
AJ001815.1 - 12 - 13 1360 69
AK225600.1 - 12 - 13 1186 69
AK299011.1 - 12 - 13 1182 69
AK307654.1 - 12 - 13 1186 69
BC062574.1 - 10 - 11 977 69
BX571745.1 - 11 - 12 2765 69

View Full Product Details