Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- BMP8B
- Entrez Gene ID
-
656
- Gene Name
- bone morphogenetic protein 8b
- Gene Aliases
-
BMP8, OP2
- Chromosome Location
-
Chr.1: 39757182 - 39788861 on Build GRCh38
- Gene Symbol
- BMP8A
- Entrez Gene ID
-
353500
- Gene Name
- bone morphogenetic protein 8a
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 39491626 - 39529869 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_181809.3 |
NP_861525.2 |
3 - 4 |
1018 |
110 |
View Full Product Details