The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000024.5 | NP_000015.1 | 1 - 1 | 778 | 65 |
GenBank mRNA | AK223025.1 | - | 1 - 1 | 741 | 65 |
AK313151.1 | - | 1 - 1 | 760 | 65 | |
AY136741.1 | - | 1 - 1 | 539 | 65 | |
BC012481.2 | - | 1 - 1 | 748 | 65 | |
BC063486.1 | - | 1 - 1 | 726 | 65 | |
BC073856.1 | - | 1 - 1 | 729 | 65 | |
KJ534762.1 | - | 1 - 1 | 539 | 65 | |
M15169.1 | - | 1 - 1 | 2126 | 65 | |
X04827.1 | - | 1 - 1 | 716 | 65 |