Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSC2
Entrez Gene ID
7249
Gene Name
tuberous sclerosis 2
Gene Aliases
LAM, PPP1R160, TSC4
Chromosome Location
Chr.16: 2047804 - 2088720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000548.4 NP_000539.2 3 - 4 427 78
NM_001077183.2 NP_001070651.1 3 - 4 427 78
NM_001114382.2 NP_001107854.1 3 - 4 427 78
NM_001318829.1 NP_001305758.1 2 - 3 260 78
NM_001318832.1 NP_001305761.1 3 - 4 328 78
XM_005255529.4 XP_005255586.2 3 - 4 314 78
XM_005255531.4 XP_005255588.2 3 - 4 314 78
XM_011522636.2 XP_011520938.1 3 - 4 314 78
XM_011522637.2 XP_011520939.1 3 - 4 314 78
XM_011522639.2 XP_011520941.1 3 - 4 314 78
XM_011522640.2 XP_011520942.1 3 - 4 314 78
XM_017023615.1 XP_016879104.1 3 - 4 314 78
XM_017023616.1 XP_016879105.1 3 - 4 314 78
XM_017023618.1 XP_016879107.1 3 - 4 314 78
GenBank mRNA AB210000.1 - 3 - 4 333 78
AK294548.1 - 3 - 4 325 78
AK295672.1 - 2 - 3 147 78
AK295728.1 - 3 - 4 328 78
BC150300.1 - 3 - 4 302 78
DA316794.1 - 3 - 4 342 78
DC402106.1 - 3 - 4 429 78
KJ534954.1 - 2 - 3 227 78
KJ534955.1 - 2 - 3 227 78
KJ534956.1 - 2 - 3 227 78
KJ534957.1 - 2 - 3 227 78
KJ534958.1 - 2 - 3 227 78
KJ534982.1 - 2 - 3 227 78
KJ534983.1 - 2 - 3 227 78
KJ534984.1 - 2 - 3 227 78
KJ534985.1 - 2 - 3 227 78
KJ535011.1 - 2 - 3 227 78
KJ535022.1 - 2 - 3 227 78
KJ535038.1 - 2 - 3 227 78
KJ535039.1 - 2 - 3 227 78
KJ535051.1 - 2 - 3 227 78
KJ535091.1 - 2 - 3 227 78
X75621.1 - 2 - 3 245 78

View Full Product Details