The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130831.1 | NP_001124303.1 | 11 - 12 | 1110 | 59 |
NM_003644.2 | NP_003635.2 | 7 - 8 | 859 | 59 | |
NM_201432.1 | NP_958836.1 | 11 - 12 | 1267 | 59 | |
NM_201433.1 | NP_958839.1 | 11 - 12 | 1297 | 59 | |
XM_005256831.3 | XP_005256888.1 | 12 - 13 | 1208 | 59 | |
XM_005256832.3 | XP_005256889.1 | 11 - 12 | 1078 | 59 | |
XM_005256833.3 | XP_005256890.1 | 10 - 11 | 1185 | 59 | |
XM_005256834.3 | XP_005256891.1 | 10 - 11 | 1011 | 59 | |
XM_005256835.3 | XP_005256892.1 | 10 - 11 | 955 | 59 | |
XM_011524044.2 | XP_011522346.1 | 11 - 12 | 1102 | 59 | |
XM_017025239.1 | XP_016880728.1 | 11 - 12 | 1131 | 59 | |
XM_017025240.1 | XP_016880729.1 | 10 - 11 | 914 | 59 | |
XM_017025241.1 | XP_016880730.1 | 10 - 11 | 890 | 59 | |
XM_017025242.1 | XP_016880731.1 | 10 - 11 | 1064 | 59 | |
GenBank mRNA | AB007854.1 | - | 11 - 12 | 1065 | 59 |
AB209030.1 | - | 10 - 11 | 954 | 59 | |
AJ224876.1 | - | 7 - 8 | 882 | 59 | |
AK057761.1 | - | 8 - 9 | 857 | 59 | |
AK292987.1 | - | 11 - 12 | 1083 | 59 | |
AK293755.1 | - | 7 - 8 | 690 | 59 | |
AK293834.1 | - | 5 - 6 | 668 | 59 | |
AK294829.1 | - | 10 - 11 | 1058 | 59 | |
AK309722.1 | - | 11 - 12 | 1118 | 59 | |
AK315162.1 | - | 7 - 8 | 872 | 59 | |
AY327406.1 | - | 11 - 12 | 1297 | 59 | |
BC001152.1 | - | 11 - 12 | 1090 | 59 | |
BC006454.1 | - | 11 - 12 | 1267 | 59 | |
BC113835.1 | - | 1 - 2 | 47 | 59 | |
BT006891.1 | - | 10 - 11 | 944 | 59 |