The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004677.2 | NP_004668.2 | 1 - 1 | 198 | 79 |
GenBank mRNA | AF000997.1 | - | 1 - 1 | 198 | 79 |
BC132883.1 | - | 1 - 1 | 70 | 79 | |
BC132885.1 | - | 1 - 1 | 70 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002906.1 | NP_001002906.1 | 1 - 1 | 198 | 79 |
GenBank mRNA | BC132901.1 | - | 1 - 1 | 70 | 79 |
BC132903.1 | - | 1 - 1 | 70 | 79 |