The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130865.2 | NP_001124337.1 | 3 - 4 | 859 | 98 |
NM_006557.6 | NP_006548.1 | 2 - 3 | 1004 | 98 | |
NM_181872.4 | NP_870987.2 | 2 - 3 | 730 | 98 | |
XM_011517687.1 | XP_011515989.1 | 4 - 5 | 3278 | 98 | |
XM_011517690.2 | XP_011515992.1 | 3 - 4 | 1358 | 98 | |
XM_017014213.1 | XP_016869702.1 | 4 - 5 | 1680 | 98 | |
XM_017014214.1 | XP_016869703.1 | 3 - 4 | 1355 | 98 | |
XM_017014215.1 | XP_016869704.1 | 2 - 3 | 1199 | 98 | |
XM_017014216.1 | XP_016869705.1 | 2 - 3 | 1492 | 98 | |
GenBank mRNA | AF130729.1 | - | 2 - 3 | 749 | 98 |
AF284223.1 | - | 1 - 2 | 530 | 98 | |
AF284224.1 | - | 1 - 2 | 530 | 98 | |
AF284225.1 | - | 1 - 2 | 530 | 98 | |
AK290794.1 | - | 2 - 3 | 1199 | 98 | |
BC030204.1 | - | 3 - 4 | 859 | 98 | |
BC136493.1 | - | 1 - 2 | 609 | 98 | |
Y19052.1 | - | 2 - 3 | 984 | 98 |