The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018057.1 | NP_001018067.1 | 1 - 2 | 272 | 83 |
NM_013253.4 | NP_037385.2 | 2 - 3 | 440 | 83 | |
NM_015881.5 | NP_056965.3 | 2 - 3 | 454 | 83 | |
XM_006718178.2 | XP_006718241.1 | 2 - 3 | 267 | 83 | |
XM_017017553.1 | XP_016873042.1 | 2 - 3 | 401 | 83 | |
XM_017017554.1 | XP_016873043.1 | 2 - 3 | 419 | 83 | |
XM_017017555.1 | XP_016873044.1 | 2 - 3 | 267 | 83 | |
GenBank mRNA | AB033421.1 | - | 1 - 2 | 233 | 83 |
AB034203.1 | - | 2 - 3 | 412 | 83 | |
AB057591.1 | - | 2 - 3 | 440 | 83 | |
AF177396.1 | - | 1 - 2 | 252 | 83 | |
AK090952.1 | - | - | 359 | 83 | |
AK092979.1 | - | 2 - 3 | 231 | 83 | |
AK225156.1 | - | 1 - 2 | 224 | 83 | |
AK289897.1 | - | 2 - 3 | 281 | 83 | |
AK295441.1 | - | 1 - 2 | 216 | 83 | |
AK295539.1 | - | 2 - 3 | 375 | 83 | |
AL535720.3 | - | 1 - 2 | 271 | 82 | |
AY358378.1 | - | 2 - 3 | 367 | 83 | |
AY587550.1 | - | 1 - 2 | 215 | 83 | |
BC007660.2 | - | 2 - 3 | 453 | 83 | |
CB156451.1 | - | 2 - 3 | 427 | 83 |