The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013423.2 | NP_038267.1 | 1 - 2 | 292 | 60 |
GenBank mRNA | AF177663.1 | - | 1 - 2 | 292 | 60 |