The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017977.2 | NP_001017977.1 | 11 - 12 | 1852 | 84 |
NM_001198956.1 | NP_001185885.1 | 13 - 14 | 2083 | 84 | |
NM_001198957.1 | NP_001185886.1 | 12 - 13 | 1990 | 84 | |
NM_018442.3 | NP_060912.2 | 12 - 13 | 1912 | 84 | |
XM_005245331.4 | XP_005245388.1 | 13 - 14 | 2024 | 84 | |
XM_005245332.4 | XP_005245389.1 | 12 - 13 | 1853 | 84 | |
XM_005245333.4 | XP_005245390.1 | 11 - 12 | 1793 | 84 | |
XM_017001779.1 | XP_016857268.1 | 13 - 14 | 1896 | 84 | |
XM_017001780.1 | XP_016857269.1 | 12 - 13 | 1661 | 84 | |
XM_017001781.1 | XP_016857270.1 | 11 - 12 | 1651 | 84 | |
GenBank mRNA | AF116725.1 | - | 13 - 14 | 1824 | 84 |
AF124434.1 | - | 10 - 11 | 1347 | 84 | |
AF150734.1 | - | 9 - 10 | 1212 | 84 | |
AK226181.1 | - | 11 - 12 | 1703 | 84 | |
AK297847.1 | - | 12 - 13 | 1850 | 84 | |
AL136738.1 | - | 4 - 5 | 497 | 84 | |
AY766164.1 | - | 11 - 12 | 1592 | 84 | |
BC025262.1 | - | 3 - 4 | 515 | 84 | |
DQ768089.1 | - | 11 - 12 | 1516 | 84 |