The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022149.4 | NP_071432.2 | 1 - 1 | 858 | 69 |
GenBank mRNA | AF295378.1 | - | 1 - 1 | 810 | 69 |
AF320910.1 | - | 1 - 1 | 729 | 69 | |
BC010056.2 | - | 1 - 1 | 815 | 69 | |
BQ058163.1 | - | 1 - 1 | 464 | 69 |