The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003097.4 | NP_003088.1 | 3 - 4 | 420 | 74 |
NM_022805.3 | NP_073716.1 | 5 - 6 | 615 | 74 | |
NM_022806.3 | NP_073717.1 | 5 - 6 | 625 | 74 | |
NM_022807.3 | NP_073718.1 | 6 - 7 | 760 | 74 | |
NM_022808.3 | NP_073719.1 | 5 - 6 | 652 | 74 | |
GenBank mRNA | AK222633.1 | - | 5 - 6 | 575 | 74 |
BC003180.1 | - | 3 - 4 | 326 | 74 | |
BC018809.2 | - | 3 - 4 | 315 | 74 | |
BC024777.1 | - | 4 - 5 | 448 | 74 | |
BC025178.1 | - | 3 - 4 | 292 | 74 | |
DA131564.1 | - | 5 - 6 | 569 | 74 | |
J04615.1 | - | 3 - 4 | 325 | 74 | |
L80004.1 | - | 5 - 6 | 604 | 74 | |
L80005.1 | - | 6 - 7 | 750 | 74 | |
U41303.1 | - | 3 - 4 | 325 | 74 | |
X15892.1 | - | 1 - 2 | 202 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005678.4 | NP_005669.2 | 3 - 4 | 420 | 74 |
GenBank mRNA | AF101044.1 | - | 3 - 4 | 325 | 74 |
AK098563.1 | - | 3 - 4 | 324 | 74 | |
DA134019.1 | - | 3 - 4 | 390 | 74 |