The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321881.1 | NP_001308810.1 | 18 - 19 | 1844 | 86 |
NM_001321882.1 | NP_001308811.1 | 18 - 19 | 2062 | 86 | |
NM_024635.3 | NP_078911.3 | 18 - 19 | 1848 | 86 | |
XM_005252127.3 | XP_005252184.1 | 18 - 19 | 1919 | 86 | |
XM_011518903.2 | XP_011517205.1 | 15 - 16 | 1651 | 86 | |
XM_011518904.2 | XP_011517206.1 | 12 - 13 | 1337 | 86 | |
XM_017015012.1 | XP_016870501.1 | 19 - 20 | 1907 | 86 | |
GenBank mRNA | AK026296.1 | - | 2 - 3 | 183 | 86 |
AK292239.1 | - | 18 - 19 | 1848 | 86 | |
BC117427.1 | - | 18 - 19 | 1769 | 86 | |
BC117429.1 | - | 18 - 19 | 1765 | 86 |