The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025077.3 | NP_079353.3 | 1 - 2 | 634 | 66 |
XM_011540569.2 | XP_011538871.1 | 1 - 2 | 847 | 66 | |
GenBank mRNA | AK024011.1 | - | 1 - 2 | 155 | 66 |
AY169960.1 | - | 1 - 2 | 631 | 66 | |
BC009364.2 | - | 1 - 2 | 68 | 66 |