Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFB2
Entrez Gene ID
80341
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2
Chromosome Location
Chr.20: 33007578 - 33023709 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025227.2 NP_079503.1 2 - 3 322 99
GenBank mRNA AF465765.1 - 1 - 2 105 99
AK027068.1 - 2 - 3 299 99
AK172819.1 - 2 - 3 301 99
AK304064.1 - 2 - 3 227 99
AY358723.1 - 2 - 3 230 99
BC034415.1 - 2 - 3 176 99
BP333980.1 - 2 - 3 321 99

View Full Product Details