The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018906.2 | NP_061729.1 | 1 - 1 | 607 | 83 |
NM_031497.1 | NP_113685.1 | 1 - 1 | 607 | 83 | |
GenBank mRNA | AF152311.1 | - | 1 - 1 | 607 | 83 |
AF152481.1 | - | 1 - 1 | 607 | 83 | |
BC069064.1 | - | 1 - 1 | 607 | 83 | |
BC129807.1 | - | 1 - 1 | 607 | 83 |