The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018926.2 | NP_061749.1 | 1 - 1 | 916 | 99 |
NM_032100.1 | NP_115271.1 | 1 - 1 | 916 | 99 | |
GenBank mRNA | AF152335.1 | - | 1 - 1 | 916 | 99 |
AF152522.1 | - | 1 - 1 | 916 | 99 |